EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Store-operated Ca(2+) entry in rhabdomyosarcoma cells.
PMID 27291153
Abstract
Rhabdomyosarcoma (RMS), the most common pediatric soft tissue sarcoma, has an intrinsic or early-acquisition of resistance to chemo- and radiation therapy. Molecular determinants pivotal for RMS migration, metastatic invasion, cell proliferation, and survival are incompletely identified. Migration and cell proliferation were shown to correlate with cytosolic Ca(2+) activity ([Ca(2+)]i). Store-operated Ca(2+)-entry (SOCE) that increases intracellular [Ca(2+)] is accomplished by Orai1, a pore-forming ion channel unit, the expression of which is stimulated by the transcription factor NFκB. The present study explored the expression of Orai1 and its regulators STIM1 and NFκB in human rhabdomyosarcoma cell lines and analyzed their impact on cell proliferation and migration. For the study human rhabdomyosarcoma cell lines RD (embryonal) and RH30 (alveolar) were analyzed for Orai1, STIM1, and NFκB transcription by RT-PCR and their corresponding proteins in Western blot. [Ca(2+)]i was detected via Fura-2 fluorescence and SOCE - resulting from [Ca(2+)]i increase following store depletion with extracellular Ca(2+) removal and inhibition of the sarcoendoplasmatic reticular Ca(2+) ATPase - detected with thapsigargin. Cell migration was analyzed in transwell and mitotic cell death with the clonogenic assay. In summary, Orai1, STIM1, and NFκB are expressed in embryonal (RD) and alveolar (RH30) rhabdomyosarcoma. SOCE inhibitor BTP2, Orai1 inhibitor 2-APB, or NFκB inhibitor wogonin virtually abrogated (BTP2, 2-APB) or significantly reduced (wogonin) SOCE. Moreover, SOCE inhibitors 2-APB and BTP2 and wogonin significantly inhibited migration and proliferation of both, RD and RH30xa0cells. These results suggest that Orai1 signaling is involved in SOCE into rhabdomyosarcoma cells thus contributing to migration, invasion and proliferation.