EMAIL THIS PAGE TO A FRIEND
Journal of pineal research
The impaired myocardial ischemic tolerance in adult offspring of diabetic pregnancy is restored by maternal melatonin treatment.
PMID 27299979
Abstract
Diabetic pregnancy, with ever increasing prevalence, adversely affects embryogenesis and increases vasculometabolic disorder risks in adult offspring. However, it remains poorly understood whether maternal diabetes increases the offspring's susceptibility to heart injuries in adulthood. In this study, we observed that cardiac function and structure were comparable between adult offspring born to diabetic mice and their counterparts born to nondiabetic mice at baseline. However, in response to myocardial ischemia/reperfusion (MIR), diabetic mother offspring exhibited augmented infarct size, cardiac dysfunction, and myocardial apoptosis compared with control, in association with exaggerated activation of mitochondria- and endoplasmic reticulum (ER) stress-mediated apoptosis pathways and oxidative stress. Molecular analysis showed that the impaired myocardial ischemic tolerance in diabetic mother offspring was mainly attributable to blunted cardiac insulin receptor substrate (IRS)-1/Akt signaling. Furthermore, the effect of maternal melatonin administration on offspring's response to MIR was determined, and the results indicated that melatonin treatment in diabetic dams during pregnancy significantly improved the tolerance to MIR injury in their offspring, via restoring cardiac IRS-1/Akt signaling. Taken together, these data suggest that maternal diabetes predisposes offspring to augmented MIR injury in adulthood, and maternal melatonin supplementation during diabetic pregnancy may hold promise for improving myocardial ischemic tolerance in the offspring.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
861650
(+)-Biotin 4-nitrophenyl ester, 98%
C16H19N3O5S