EMAIL THIS PAGE TO A FRIEND
Biomaterials
A high affinity kidney targeting by chitobionic acid-conjugated polysorbitol gene transporter alleviates unilateral ureteral obstruction in rats.
PMID 27318934
Abstract
Aside from kidney transplantation - a procedure which is exceedingly dependent on donor-match and availability leading to excessive costs - there are currently no permanent treatments available which reverse kidney injury and failure. However, kidney-specific targeted gene therapy has outstanding potential to treat kidney-related dysfunction. Herein we report a novel kidney-specific targeted gene delivery system developed through the conjugation of chitobionic acid (CBA) to a polysorbitol gene transporter (PSGT) synthesized from sorbitol diacrylate and low molecular weight polyethylenimine (PEI) carrying hepatocyte growth factor (HGF) gene to alleviate unilateral ureteral obstruction (UUO) in rats. CBA-PSGT performed exceptionally well for targeted delivery of HGF to kidney tissues compared to its non-targeted counterparts (Pxa0