EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Stable fluorescence conjugation of ZnO nanoparticles and their size dependent cellular uptake.
PMID 27323344
Abstract
We evaluated size dependent cellular uptake of ZnO nanoparticles utilizing stably introduced Cy5.5, which emits long-wavelength fluorescence. Through (3-aminopropyl)triethoxysilane modification, ZnO nanoparticles of different sizes (20 and 70nm) were functionalized with amine moiety, which was further reacted with Cy5.5-N-hydroxylsuccinimide ester to make covalently conjugated Cy5.5 dye on ZnO nanoparticles. Field emission-scanning electron microscopic images revealed that average particle size as well as particle morphology of ZnO nanoparticles were not altered by Cy5.5 conjugation. Zeta potential measurement confirmed that the positive surface charge of ZnO nanoparticles was well preserved after successive conjugation reactions. Based on infrared, ultraviolet-visible light and photoluminescence spectroscopies, we verify that the Cy5.5 was stably introduced to ZnO nanoparticles without serious aggregation. Surface conjugated Cy5.5 showed high stability in deionized water, phosphate buffered saline and cell culture medium, showing less than 2% of release during 85h. Confocal microscopy and fluorescence-activated cell sorting analysis demonstrated that smaller ZnO nanoparticles were more taken up in greater quantities by HaCaT cells. Moreover, systematic study on cellular uptake pathway showed that smaller ZnO nanoparticles were internalized into cells mainly by clathrin-mediated endocytosis, while larger ZnO nanoparticles entered cells via several pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
244511
Toluene, anhydrous, 99.8%
C7H8