EMAIL THIS PAGE TO A FRIEND
Cell death and differentiation
A siRNA screen reveals the prosurvival effect of protein kinase A activation in conditions of unresolved endoplasmic reticulum stress.
PMID 27341185
Abstract
The endoplasmic reticulum (ER) has a crucial role in the proper folding of proteins that are synthesized in the secretory pathway. Physiological and pathological conditions can induce accumulation of mis- or unfolded proteins in the ER lumen and thereby generate a state of cellular stress known as ER stress. The unfolded protein response aims at restoring protein-folding homeostasis, but turns into a toxic signal when ER stress is too severe or prolonged. ER stress-induced cellular dysfunction and death is associated with several human diseases, but the molecular mechanisms regulating death under unresolved ER stress are still unclear. We performed a siRNA-based screen to identify new regulators of ER stress-induced death and found that repression of the Carney complex-associated protein PRKAR1A specifically protected the cells from ER stress-induced apoptosis, and not from apoptosis induced by etoposide or TNF. We demonstrate that the protection results from PKA activation and associate it, at least in part, with the phosphorylation-mediated inhibition of the PKA substrate Drp1 (dynamin-related protein 1). Our results therefore provide new information on the complex regulation of cellular death under ER stress conditions and bring new insights on the conditions that regulate the pro- versus anti-death functions of PKA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7765
Tunicamycin from Streptomyces sp.