EMAIL THIS PAGE TO A FRIEND
General and comparative endocrinology
Sphingosine-1-phosphate, regulated by FSH and VEGF, stimulates granulosa cell proliferation.
PMID 27342378
Abstract
Sphingosine-1-phosphate (S1P) is a bioactive polar sphingolipid which stimulates proliferation, growth and survival in various cell types. In the ovary S1P has been shown protect the granulosa cells and oocytes from insults such as oxidative stress and radiotherapy, and S1P concentrations are greater in healthy than atretic large follicles. Hence, we postulate that S1P is fundamental in follicle development and that it is activated in ovarian granulosa cells in response to FSH and VEGF. To test this hypothesis we set out: i) to evaluate the effect of FSH and VEGF on S1P synthesis in cultured bovine granulosa cells and ii) to analyse the effect of S1P on proliferation and survival of bovine granulosa cells in vitro. Seventy five thousand bovine granulosa cells from healthy medium-sized (4-7mm) follicles were cultured in 96-well plates in McCoy's 5a medium containing 10ng/mL of insulin and 1ng/mL of LR-IGF-I at 37°C in a 5% CO2/air atmosphere at 37°C. Granulosa cell production of S1P was tested in response to treatment with FSH (0, 0.1, 1 and 10ng/mL) and VEGF (0, 0.01, 0.1, 1, 10 and 100ng/mL) and measured by HPLC. Granulosa cells produced S1P at 48 and 96h, with the maximum production observed with 1ng/mL of FSH. Likewise, 0.01ng/mL of VEGF stimulated S1P production at 48, but not 96h of culture. Further, the granulosa cell expression of sphingosine kinase-1 (SK1), responsible for S1P synthesis, was demonstrated by Western blot after 48h of culture. FSH increased the expression of phosphorylated SK1 (P<0.05) and the addition of a SK1 inhibitor reduced the constitutive and FSH-stimulated S1P synthesis (P<0.05). Sphingosine-1-phosphate had a biphasic effect on granulosa cell number after culture. At low concentration S1P (0.1μM) increased granulosa cell number after 48h of culture (P<0.05) and the proportion of cells in the G2 and M phase of the cell cycle (P<0.05), whereas higher concentrations decreased cell number (10μM; P<0.05) by an increase (P<0.05) in the proportion of cells in apoptosis (hypodiploid cells). In addition, treatment with SK-178 suppressed the FSH- and VEGF-stimulated rise of the granulosa cells number (P<0.05). Interestingly, the effect of 0.1μM S1P on granulosa cell number and their proportion in G2/M phases is similar to that observed with 1ng/mL FSH. The results of this study are the first to demonstrate sphingosine-1-phosphate (S1P) synthesis in granulosa cells under the control of FSH and VEGF. The later achieved through the regulation of sphingosine kinase 1 expression. This S1P augments the proportion of cells in the G2/M phase of the cell cycle that translates in increased granulosa cell proliferation.