EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie. International edition in English
Nickel-Catalyzed Cross-Coupling of Redox-Active Esters with Boronic Acids.
PMID 27380912
Abstract
A transformation analogous in simplicity and functional group tolerance to the venerable Suzuki cross-coupling between alkyl-carboxylic acids and boronic acids is described. This Ni-catalyzed reaction relies upon the activation of alkyl carboxylic acids as their redox-active ester derivatives, specifically N-hydroxy-tetrachlorophthalimide (TCNHPI), and proceeds in a practical and scalable fashion. The inexpensive nature of the reaction components (NiCl2 ⋅6 H2 O-$9.5 mol(-1) , Et3 N) coupled to the virtually unlimited commercial catalog of available starting materials bodes well for its rapid adoption. © 2016 WILEY-VCH Verlag GmbH & Co. KGaA, Weinheim.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ALD00564
N-Hydroxytetrachlorophthalimide
C8HCl4NO3
Set your institution to read full text papers when available.