EMAIL THIS PAGE TO A FRIEND
Journal of dental research
DPSCs from Inflamed Pulp Modulate Macrophage Function via the TNF-α/IDO Axis.
PMID 27384335
Abstract
Human dental pulp stem cells (DPSCs) can be isolated from inflamed pulp derived from carious teeth with symptomatic irreversible pulpitis (I-DPSCs), which possess stemness and multidifferentiation potentials similar to DPSCs from healthy pulp. Since macrophages-essential cell players of the pulpal innate immunity-can regulate pulpal inflammation and repair, the authors investigated the immunomodulatory effects of DPSCs/I-DPSCs on macrophage functions and their underlying mechanisms. Similar to DPSCs, I-DPSCs were capable of colony-forming efficiency and adipogenic and osteo/dentinogenic differentiation under in vitro induction conditions. I-DPSCs also expressed a similar phenotypic profile of mesenchymal stem cell markers, except a relatively higher level of CD146 as compared with DPSCs. Coculture of DPSCs or I-DPSCs with differentiated THP-1 cells, the human monocyte cell line, markedly suppressed tumor necrosis factor α (TNF-α) secretion in response to stimulation with lipopolysaccharides (LPS) and/or nigericin. However, unlike TNF-α, the secreted level of interleukin 1β was not affected by coculture with DPSCs or I-DPSCs. Furthermore, DPSC/I-DPSC-mediated inhibition of TNF-α secretion by macrophages was abolished by pretreatment with 1-methyl-D-tryptophan, a specific inhibitor of indoleamine-pyrrole 2,3-dioxygenase (IDO), but not by NSC-398, a specific inhibitor of COX-2, suggesting IDO as a mediator. Interestingly, IDO expression was significantly augmented in macrophages and mesenchymal stromal cells in inflamed human pulp tissues. Collectively, these findings show that I-DPSCs, similar to DPSCs, possess stem cell properties and suppress macrophage functions via the TNF-α/IDO axis, thereby providing a physiologically relevant context for their innate immunomodulatory activity in the dental pulp and their capability for pulp repair.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
452483
1-Methyl-D-tryptophan, 95%
C12H14N2O2