EMAIL THIS PAGE TO A FRIEND
Bioelectrochemistry (Amsterdam, Netherlands)
Kinetics of cytochrome c oxidase from R. sphaeroides initiated by direct electron transfer followed by tr-SEIRAS.
PMID 27398977
Abstract
Time-resolved surface-enhanced IR-absorption spectroscopy (tr-SEIRAS) has been performed on cytochrome c oxidase from Rhodobacter sphaeroides. The enzyme was converted electrochemically into the fully reduced state. Thereafter, in the presence of oxygen, the potential was switched to open circuit potential (OCP). Under these conditions, the enzyme is free to undergo enzymatic oxidation in the absence of an external electric field. Tr-SEIRAS was performed using the step-scan technique, triggered by periodic potential pulses switching between - 800mV and OCP. Single bands were resolved in a broad band in the amide I region using phase sensitive detection. Amplitudes of these bands were analyzed as a function of time. Time constants in the ms time scale were considered in terms of conformational changes of the protein secondary structures associated with the enzymatic turnover of the protein.