EMAIL THIS PAGE TO A FRIEND
Scientific reports
Bursaphelenchus xylophilus is killed by homologues of 2-(1-undecyloxy)-1-ethanol.
PMID 27403600
Abstract
2-(1-Undecyloxy)-1-ethanol, monochamol, is a male-produced aggregation pheromone of the Monochamus species, which are efficient vectors of the pine wood nematode (PWN), Bursaphelenchus xylophilus, which cause devastating damage to pines worldwide. The nematicidal activity of synthetic monochamol and its homologues (ROEtOH: R = C7-C13) were investigated to find potential alternatives to the currently used PWN control agents abamectin and emamectin. Compounds with C7-C13 chain length alkyl groups exhibited 100% nematicidal activity at a concentration of 1000 mg/L. At a concentration of 100 mg/L, 2-(1-nonyloxy)-1-ethanol (C9OEtOH), 2-(1-decyloxy)-1-ethanol (C10OEtOH), 2-(1-undecyloxy)-1-ethanol (C11OEtOH), and 2-(1-dodecyloxy)-1-ethanol (C12OEtOH) showed 100% nematicidal activity, but the others showed weaker activities. C11OEtOH showed similar nematicidal activity to abamectin in terms of LD90 values, which were 13.30 and 12.53 mg/L, respectively. However, C9OEtOH, C10OEtOH, and C12OEtOH (LC90 values: 53.63, 38.18, and 46.68 mg/L, respectively) were less effective than C11OEtOH and abamectin. These results indicate that monochamol could be an effective alternative agent against PWN. The relationship of insecticidal and nematicidal activity to different carbon chain lengths in compounds is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U1001
1-Undecanol, 99%
C11H24O