EMAIL THIS PAGE TO A FRIEND
Applied spectroscopy
Two Mechanisms of Tip Enhancement of Raman Scattering by Protein Aggregates.
PMID 27407009
Abstract
Tip-enhanced Raman spectroscopy (TERS) is a powerful tool for probing the surface of biological species with nanometer spatial resolution. Here, we report the TER spectra of an individual insulin fibril, the protein cast film and a short peptide (LVEALYL) microcrystal mimicking the fibril core. Two different types of TER spectra were acquired depending on the "roughness" of the probed surface at the molecular level. A fully reproducible, low-intensity, normal Raman-type spectrum was characteristic of the top flat surface of the microcrystal while highly variable, higher intensity TER spectra were obtained for the edges of the microcrystal, cast film, and fibril. As a result, two tip enhancement mechanisms of Raman scattering, long- and short-range, were proposed by analogy with the physical and chemical enhancement mechanisms, respectively, known for surface-enhanced Raman spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5500
Insulin from bovine pancreas, ≥25 USP units/mg (HPLC), powder
C254H377N65O75S6