EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
Immunoglobulin G elution in protein A chromatography employing the method of chromatofocusing for reducing the co-elution of impurities.
PMID 27425244
Abstract
Purification processes for monoclonal Immunoglobulin G (IgG) typically employ protein A chromatography as a capture step to remove most of the impurities. One major concern of the post-protein A chromatography processes is the co-elution of some of the host cell proteins (HCPs) with IgG in the capture step. In this work, a novel method for IgG elution in protein A chromatography that reduces the co-elution of HCPs is presented where a two-step pH gradient is self-formed inside a protein A chromatography column. The complexities involved in using an internally produced pH gradient in a protein A chromatography column employing adsorbed buffering species are discussed though equation-based modeling. Under the conditions employed, ELISA assays show a 60% reduction in the HCPs co-eluting with the IgG fraction when using the method as compared to conventional protein A elution without affecting the IgG yield. Evidence is also obtained which indicates that the amount of leached protein A present in free solution in the purified product is reduced by the new method. Biotechnol. Bioeng. 2017;114: 154-162. © 2016 Wiley Periodicals, Inc.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8883
4-Methylumbelliferyl phosphate, phosphatase substrate
C10H9O6P