EMAIL THIS PAGE TO A FRIEND
European journal of nuclear medicine and molecular imaging
Relationships between cardiac innervation/perfusion imbalance and ventricular arrhythmias: impact on invasive electrophysiological parameters and ablation procedures.
PMID 27430947
Abstract
To assess the relationship between regional myocardial perfusion and sympathetic innervation parameters at myocardial scintigraphy and intra-cavitary electrophysiological data in patients with ventricular arrhythmias (VA) submitted to invasive electrophysiological study and ablation procedure. Sixteen subjects underwent invasive electrophysiological study with electroanatomical mapping (EAM) followed by trans-catheter ablations of VA. Before ablation all patients were studied with a combined evaluation of regional myocardial perfusion and sympathetic innervation by means of tomographic (99m)Tc-tetrofosmin and (123)I- metaiodobenzylguanidine cadmium-zinc-telluride (CZT) scintigraphies, respectively. Off-line spatial co-registration of CZT perfusion and innervation data with the three-dimensional EAM reconstruction was performed in every patient. CZT revealed the presence of myocardial scar in 55 (20xa0%) segments. Of the viable myocardial segments, 131 (60xa0%) presented a preserved adrenergic innervation, while 86 (40xa0%) showed a significantly depressed innervation (i.e. innervation/perfusion mismatch). On EAM, the invasively measured intra-cavitary voltage was significantly lower in scarred segments than in viable ones (1.7 ± 1.5xa0mV vs. 4.0 ± 2.2xa0mV, P < 0.001). Interestingly, among the viable segments, those showing an innervation/perfusion mismatch presented a significantly lower intra-cavitary voltage than those with preserved innervation (1.9 ± 2.5xa0mV vs. 4.7 ± 2.3xa0mV, P < 0.001). Intra-cardiac ablation was performed in 63 (23xa0%) segments. On multivariate analysis, after correction for scar burden, the segments showing an innervation/perfusion mismatch remained the most frequent ablation targets (OR 5.6, 95xa0% CI 1.5-20.8; P = 0.009). In patients with VA, intra-cavitary electrical abnormalities frequently originate at the level of viable myocardial segments with depressed sympathetic innervation that frequently represents the ultimate ablation target.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
716707
Cadmium zinc telluride, 99.9999% trace metals basis
Cd0.9Zn0.1Te