EMAIL THIS PAGE TO A FRIEND
Macromolecular rapid communications
Ratiometric Fluorescent pH Probes Based on Glycopolymers.
PMID 27439338
Abstract
Effectively detecting pH changes plays a critical role in exploring cellular functions and determining physiological and pathological processes. A novel ratiometric pH probe based on a glycopolymer, armored with properties of serum-stability, tumor-targeting, and pH monitoring, is designed. Random copolymers of 2-(methacrylamido) glucopyranose and fluorescein O-methacrylate are first synthesized by reversible addition fragmentation chain transfer polymerization. Acryloxyethyl thiocarbamoyl rhodamine B is then attached to the polymer chain to prepare ratiometric fluorescent pH probes via a thiol-ene reaction. The synthesized polymeric probes are characterized by NMR, gel permeation chromatography, UV-vis spectroscopy, and transmission electron microscopy, and the fluorescence responses are examined in phosphate buffer at different pHs. The cytotoxicity and confocal imaging experiments of the probes are detected using HeLa cells, demonstrating a low toxicity and superior biocompatibility for detecting pH changes in bioapplications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
733342
9-Fluorenyl methacrylate, 97%
C17H14O2