EMAIL THIS PAGE TO A FRIEND
Medicinal chemistry (Shariqah (United Arab Emirates))
The Role of Size in Development of Mucosal Liposome-Lipopeptide Vaccine Candidates Against Group A Streptococcus.
PMID 27449794
Abstract
Group A streptococcus (GAS) is an exclusively human pathogenic bacteria. A delay in treatment of GAS infection often lead to severe diseases such as rheumatic heart disease which attributes to hundreds of thousands deaths annually. For the past few decades, the quest for a commercial GAS vaccine has been futile. Currently one of the most investigated strategies to develop vaccine against GAS includes the use of conserved epitopes from major virulent factor of GAS, M-protein. In this study, cationic liposomes of various sizes (70 nm to 1000 nm) were prepared with dimethyldioctadecylammonium bromide (DDAB) encapsulating lipopeptide bearing M-protein derived B-cell epitope (J14). Smaller liposomes induced higher antibody titres, though the differences between groups were not statistically significant. Nonetheless, all mice which were immunized with liposome-lipopeptide delivery system elicited high levels of systemic (IgG) and mucosal antibodies (IgA), which were discernably higher than those induced with the help of commercial adjuvant (cholera toxin B subunit).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
382310
Didecyldimethylammonium bromide, 98%
C22H48BrN