EMAIL THIS PAGE TO A FRIEND
Oncotarget
Over-expression of ASIC1a promotes proliferation via activation of the β-catenin/LEF-TCF axis and is associated with disease outcome in liver cancer.
PMID 27462920
Abstract
Acid-sensing ion channels 1a (ASIC1a) has been reported to promote migration and invasion in liver cancer. However, the clinical significance and molecular mechanism of ASIC1a in liver cancer remain unknown. In the study, we found that ASIC1a is frequently up-regulated in liver cancer tissues. The over-expression of ASIC1a is associated with advanced clinical stage and poor prognosis. The pro-proliferative of ASIC1a is pH dependent. Knockout of ASIC1a by CRISPR/CAS9 inhibited liver cancer cell proliferation and tumorigenicity in vitro and in vivo through β-catenin degradation and LEF-TCF inactivation. Our results indicated a potential diagnostic marker and chemotherapeutic target for liver cancer.