EMAIL THIS PAGE TO A FRIEND
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals
Tissue-based quantitative proteome analysis of human hepatocellular carcinoma using tandem mass tags.
PMID 27467182
Abstract
Human hepatocellular carcinoma (HCC) is a severe malignant disease, and accurate and reliable diagnostic markers are still needed. This study was aimed for the discovery of novel marker candidates by quantitative proteomics. Proteomic differences between HCC and nontumorous liver tissue were studied by mass spectrometry. Among several significantly upregulated proteins, translocator protein 18 (TSPO) and Ras-related protein Rab-1A (RAB1A) were selected for verification by immunohistochemistry in an independent cohort. For RAB1A, a high accuracy for the discrimination of HCC and nontumorous liver tissue was observed. RAB1A was verified to be a potent biomarker candidate for HCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86600
Tetraethylammonium borohydride, technical, ≥95% (T)
C8H24BN