EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Concentrations and identification of culturable airborne fungi in underground stations of the Seoul metro.
PMID 27473614
Abstract
The purpose of this study was to measure the culturable airborne fungi (CAF) concentrations in the underground subway stations of Seoul, Korea at two time points. This study measured the CAF concentrations in enclosed environments at 16 underground stations of the Seoul Metro in 2006 and 2013 and investigated the effects of various environmental factors, including the presence of platform screen doors, temperature, relative humidity, and number of passengers. CAF concentrations at the stations in 2006 were significantly higher than that at the same stations in 2013 (pxa0