EMAIL THIS PAGE TO A FRIEND
Immunology
The contribution of the citrate pathway to oxidative stress in Down syndrome.
PMID 27502741
Abstract
Inflammatory conditions and oxidative stress have a crucial role in Down syndrome (DS). Emerging studies have also reported an altered lipid profile in the early stages of DS. Our previous works demonstrate that citrate pathway activation is required for oxygen radical production during inflammation. Here, we find up-regulation of the citrate pathway and down-regulation of carnitine/acylcarnitine carrier and carnitine palmitoyl-transferase 1 genes in cells from children with DS. Interestingly, when the citrate pathway is inhibited, we observe a reduction in oxygen radicals as well as in lipid peroxidation levels. Our preliminary findings provide evidence for a citrate pathway dysregulation, which could be related to some phenotypic traits of people with DS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6773
DL-Malic acid disodium salt, ≥95% (capillary GC)
C4H4Na2O5