EMAIL THIS PAGE TO A FRIEND
Journal of the American Society for Mass Spectrometry
Action-FRET of a Gaseous Protein.
PMID 27506208
Abstract
Mass spectrometry is an extremely powerful technique for analysis of biological molecules, in particular proteins. One aspect that has been contentious is how much native solution-phase structure is preserved upon transposition to the gas phase by soft ionization methods such as electrospray ionization. To address this question-and thus further develop mass spectrometry as a tool for structural biology-structure-sensitive techniques must be developed to probe the gas-phase conformations of proteins. Here, we report Förster resonance energy transfer (FRET) measurements on a ubiquitin mutant using specific photofragmentation as a reporter of the FRET efficiency. The FRET data is interpreted in the context of circular dichroism, molecular dynamics simulation, and ion mobility data. Both the dependence of the FRET efficiency on the charge state-where a systematic decrease is observed-and on methanol concentration are considered. In the latter case, a decrease in FRET efficiency with methanol concentration is taken as evidence that the conformational ensemble of gaseous protein cations retains a memory of the solution phase conformational ensemble upon electrospray ionization. Graphical Abstract ᅟ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
546062
Rhodamine 19 perchlorate, Dye content ~95 %
C26H26N2O3 · HClO4