EMAIL THIS PAGE TO A FRIEND
Food chemistry
Simultaneous determination of individual isothiocyanates in plant samples by HPLC-DAD-MS following SPE and derivatization with N-acetyl-l-cysteine.
PMID 27507514
Abstract
The procedure for the isothiocyanates (ITCs) determination that involves derivatization with N-acetyl-l-cysteine (NAC) and separation by HPLC was developed. Prior to derivatization, plant ITCs were isolated and purified using solid-phase extraction (SPE). The optimum conditions of derivatization are: 500μL of isopropanolic eluate obtained by SPE combined with 500μL of derivatizing reagent (0.2M NAC and 0.2M NaHCO3 in water) and reaction time of 1h at 50°C. The formed dithiocarbamates are directly analyzed by HPLC coupled with diode array detector and mass spectrometer if required. The method was validated for nine common natural ITCs. Calibration curves were linear (R(2)⩾0.991) within a wide range of concentrations and limits of detection were below 4.9nmol/mL. The recoveries were in the range of 83.3-103.7%, with relative standard deviations <5.4%. The developed method has been successfully applied to determine ITCs in broccoli, white cabbage, garden cress, radish, horseradish and papaya.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
673099
Biphenyl-4,4′-dithiol, 95%
C12H10S2