EMAIL THIS PAGE TO A FRIEND
Mutagenesis
RUNX2 mutation impairs bone remodelling of dental follicle cells and periodontal ligament cells in patients with cleidocranial dysplasia.
PMID 27509906
Abstract
RUNX2 is an important osteo-specific factor with crucial functions in bone formation and remodelling as well as resorption of teeth. Heterozygous mutation of RUNX2 can cause cleidocranial dysplasia (CCD), a systemic disease with extensive skeletal dysplasia and abnormality of tooth growth. In our study, dental follicle cells (DFCs) and periodontal ligament cells (PDLCs) were isolated, cultured and identified from one patient with CCD and compared with normal controls. This CCD patient was confirmed to have a heterozygous frameshift mutation of RUNX2 (c.514delT, p.Ser172fs) in the previous study. The results showed that the proliferation abilities of DFCs and PDLCs were both disturbed by the RUNX2 mutation in the CCD patient compared with the normal control. A co-culture system of these cells with human peripheral blood mononuclear cells was then used to investigate the effect of RUNX2 mutation on osteoclastogenesis. We found that the RUNX2 mutation in CCD reduced the expression of osteoclast-related genes, such as RUNX2, CTR, CTSK, RANKL and OPG The ability of osteoclastogenesis in DFCs and PDLCs detected by tartrate-resistant acid phosphatase staining in the co-culture system was also reduced by the RUNX2 mutation compared with the normal control. These outcomes indicate that the RUNX2 mutation disturbs the modulatory effects of DFCs and PDLCs on the differentiation of osteoclasts and osteoblasts, thereby interfering with bone remodelling. These effects may contribute in part to the pathological manifestations of retention of primary teeth and delayed eruption of permanent teeth in patients with CCD.