EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
Development & automation of a novel [(18)F]F prosthetic group, 2-[(18)F]-fluoro-3-pyridinecarboxaldehyde, and its application to an amino(oxy)-functionalised Aβ peptide.
PMID 27518217
Abstract
2-[(18)F]-Fluoro-3-pyridinecarboxaldehyde ([(18)F]FPCA) is a novel, water-soluble prosthetic group. It's radiochemistry has been developed and fully-automated for application in chemoselective radiolabelling of amino(oxy)-derivatised RI-OR2-TAT peptide, (Aoa-k)-RI-OR2-TAT, using a GE TRACERlab FX-FN. RI-OR2-TAT is a brain-penetrant, retro-inverso peptide that binds to amyloid species associated with Alzheimer's Disease. Radiolabelled (Aoa-k)-RI-OR2-TAT was reproducibly synthesised and the product of the reaction with FPCA has been fully characterised. In-vivo biodistribution of [(18)F]RI-OR2-TAT has been measured in Wistar rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
664111
2-Fluoro-3-pyridinecarboxaldehyde, 97%
C6H4FNO