EMAIL THIS PAGE TO A FRIEND
FEBS letters
Digital fluorescence imaging of fusion of influenza virus with erythrocytes.
PMID 2753145
Abstract
Fusion of influenza virus with human erythrocytes at pH 5.2 was followed by fluorescence microscopy using a cooled slow-scan CCD camera. The high sensitivity of the CCD permits repetitive digital imaging of the same cells with minimal photobleaching. The experimental conditions were such that only a small number of virus particles were adsorbed per cell. Quantitative analysis of the data indicated that for most cells only a single fusion event took place. This was, however, sufficient to cause haemolysis within 30 min at 20-22 degrees C for about 60% of cells. There was a highly variable time lag between fusion and haemolysis. The lateral diffusion coefficient of virus particles on the cell surface when bound at pH 7.4 was less than 2 x 10(-13) cm2.s-1. The technique should be of value for more detailed studies of the dynamics of viral and other membrane fusion events.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83685
Rhodamine B octadecyl ester perchlorate, suitable for fluorescence, ≥98.0% (TLC)
C46H67ClN2O7