EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Isolation, characterization and determination of trace organic impurities in FD&C red no. 40.
PMID 2753932
Abstract
The unsulfonated aromatic amine 4-nitro-p-cresidine (2-methoxy-5-methyl-4-nitrobenzenamine) was identified as an impurity in the regulated color additive FD&C Red. No. 40. The compound was isolated from the water-soluble color by extraction with chloroform, followed by transfer of the free amines to acid solution and subsequent separation by reversed-phase high-performance liquid chromatography. The 4-nitro-p-cresidine was collected and then identified by gas chromatography-mass spectrometry. The levels of 4-nitro-p-cresidine as well as p-cresidine and aniline were determined in commercial batches of FD&C Red. No. 40.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
400424
6-Hydroxy-2-naphthalenesulfonic acid sodium salt hydrate, technical grade
C10H7NaO4S · xH2O