EMAIL THIS PAGE TO A FRIEND
Journal of psychosocial oncology
Personality and psychological distress among older adult, long-term cancer survivors.
PMID 27541961
Abstract
This research examines a model of how personality (Five-Factor Model) is related to adjustment to cancer in later life in terms of the presence of continuing cancer-related worry and depression among older adult, long-term cancer survivors. Data from an NCI-funded study with 275 older adult (age 60+), long-term (5+ years) survivors of breast, prostate, and colorectal cancer were examined. Regression analyses identified neuroticism as the strongest predictor of cancer-related worry along with continuing cancer-related symptoms. For depression, three personality dimensions (neuroticism, conscientiousness, and agreeableness) were significant predictors. Findings suggest the importance of considering the central role that survivors' personality characteristics play in understanding cancer-related worries and depression. Understanding these dispositional characteristics is key for social workers and health-care practitioners in counseling survivors experiencing these common mental health effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34138
Neosolaniol solution, 100 μg/mL in acetonitrile, analytical standard
C19H26O8