EMAIL THIS PAGE TO A FRIEND
Journal of microencapsulation
Optimisation of one-step desolvation and scale-up of gelatine nanoparticle production.
PMID 27556342
Abstract
Gelatine nanoparticles (GNPs) are biodegradable and biocompatible drug delivery systems with excellent clinical performances. A two-step desolvation is commonly used for their preparation, although this methodology has several shortcomings: lack of reproducibility, small scales and low yields. A straightforward and more consistent GNP preparation approach is presented here focusing on the development of a one-step desolvation with the use of a commercially available gelatine type. Controlled stirring conditions and ultrafiltration are used to achieve large-scale production of nanoparticles of up to 2.6 g per batch. Particle size distributions are conserved and comparable to those determined for two-step desolvation on small scale. Additionally, a range of cross-linking agents is examined for their effectiveness in stabilising GNPs as an alternative to glutaraldehyde. Glyceraldehyde demonstrated outstanding properties, which led to high colloidal stability. This approach optimises the manufacturing process and the scale-up of the production capacity, providing a clear potential for future applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284556
(2-Aminoethyl)trimethylammonium chloride hydrochloride, 99%
C5H15ClN2 · HCl