EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
Chenodeoxycholic acid requires activation of EGFR, EPAC, and Ca2+ to stimulate CFTR-dependent Cl- secretion in human colonic T84 cells.
PMID 27558159
Abstract
Bile acids are known to initiate intricate signaling events in a variety of tissues, primarily in the liver and gastrointestinal tract. Of the known bile acids, only the 7α-dihydroxy species, deoxycholic acid and chenodeoxycholic acid (CDCA), and their conjugates, activate processes that stimulate epithelial Cl(-) secretion. We have previously published that CDCA acts in a rapid manner to stimulate colonic ion secretion via protein kinase A (PKA)-mediated activation of the dominant Cl(-) channel, the cystic fibrosis transmembrane conductance regulator (CFTR) (Ao M, Sarathy J, Domingue J, Alrefai WA, and Rao MC. Am J Physiol Cell Physiol 305: C447-C456, 2013); however, PKA signaling did not account for the entire CDCA response. Here we show that in human colonic T84 cells, CDCA's induction of CFTR activity, measured as changes in short-circuit current (Isc), is dependent on epidermal growth factor receptor (EGFR) activation and does not involve the bile acid receptors TGR5 or farnesoid X receptor. CDCA activation of Cl(-) secretion does not require Src, mitogen-activated protein kinases, or phosphoinositide 3-kinase downstream of EGFR but does require an increase in cytosolic Ca(2+) In addition to PKA signaling, we found that the CDCA response requires the novel involvement of the exchange protein directly activated by cAMP (EPAC). EPAC is a known hub for cAMP and Ca(2+) cross talk. Downstream of EPAC, CDCA activates Rap2, and changes in free cytosolic Ca(2+) were dependent on both EPAC and EGFR activation. This study establishes the complexity of CDCA signaling in the colonic epithelium and shows the contribution of EGFR, EPAC, and Ca(2+) in CDCA-induced activation of CFTR-dependent Cl(-) secretion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU076761 MISSION® esiRNA, esiRNA human EGFR (esiRNA1)