EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Near-Infrared Optical Imaging Noninvasively Detects Acutely Damaged Muscle.
PMID 27565039
Abstract
Muscle damage is currently assessed through methods such as muscle biopsy, serum biomarkers, functional testing, and imaging procedures, each with its own inherent limitations, and a pressing need for a safe, repeatable, inexpensive, and noninvasive modality to assess the state of muscle health remains. Our aim was to develop and assess near-infrared (NIR) optical imaging as a novel noninvasive method of detecting and quantifying muscle damage. An immobilization-reambulation model was used for inducing muscle damage and recovery in the lower hindlimbs in mice. Confirmation of muscle damage was obtained using inxa0vivo indocyanine green-enhanced NIR optical imaging, magnetic resonance imaging, and exxa0vivo tissue analysis. The soleus of the immobilized-reambulated hindlimb was found to have a greater amount of muscle damage compared to that in the contralateral nonimmobilized limb, confirmed by inxa0vivo indocyanine green-enhanced NIR optical imaging (3.86-fold increase in radiant efficiency), magnetic resonance imaging (1.41-fold increase in T2), and an exxa0vivo spectrophotometric assay of indocyanine green uptake (1.87-fold increase in normalized absorbance). Contrast-enhanced NIR optical imaging provides a sensitive, rapid, and noninvasive screening method that can be used for imaging and quantifying muscle damage and recovery inxa0vivo.