EMAIL THIS PAGE TO A FRIEND
Age (Dordrecht, Netherlands)
Tinetti mobility test is related to muscle mass and strength in non-institutionalized elderly people.
PMID 27566307
Abstract
Elderly people are characterized by a high prevalence of falls and sarcopenia. However, the relationship among Tinetti mobility test (TMT) score, a powerful tool to detect elderly people at risk of falls, and sarcopenia is still not thoroughly investigated. Thus, to determine the relationship between TMT score and muscle mass and strength, 337 elderly participants (mean age 77.1 ± 6.9xa0years) admitted to comprehensive geriatric assessment were enrolled. TMT score, muscle mass by bioimpedentiometer, and muscle strength by grip strength were evaluated. Muscle mass progressively decreased as TMT score decreased (from 15.3 ± 3.7 to 8.8 ± 1.8xa0kg/m(2); p for trend <0.001). Similarly, muscle strength decreased progressively as Tinetti score decreased (from 34.7 ± 8.0 to 23.7 ± 8.7xa0kg; p for trend 0.001). Linear regression analysis demonstrated that TMT score is linearly related with muscle mass (y = 4.5x + 0.4, r = 0.61; p < 0.01) and strength (y = 14.0x + 0.8, r = 0.53; p < 0.01). Multivariate analysis confirms the strong relationship between the TMT score and muscle mass (r = 0.48, p = 0.024) and strength (r = 0.39, p = 0.046). The present study indicates that TMT score is significantly related to muscle mass and strength in non-institutionalized elderly participants. This evidence suggests that TMT score, together with evaluation of muscle mass and strength, may identify sarcopenic elderly participants at high risk of falls.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0531
BIO-Acetoxime, ≥98% (HPLC)
C18H12BrN3O3