EMAIL THIS PAGE TO A FRIEND
ACS chemical neuroscience
Synthesis and Pharmacological Evaluation of [(11)C]Granisetron and [(18)F]Fluoropalonosetron as PET Probes for 5-HT3 Receptor Imaging.
PMID 27571447
Abstract
Serotonin-gated ionotropic 5-HT3 receptors are the major pharmacological targets for antiemetic compounds. Furthermore, they have become a focus for the treatment of irritable bowel syndrome (IBS) and there is some evidence that pharmacological modulation of 5-HT3 receptors might alleviate symptoms of other neurological disorders. Highly selective, high-affinity antagonists, such as granisetron (Kytril) and palonosetron (Aloxi), belong to a family of drugs (the "setrons") that are well established for clinical use. To enable us to better understand the actions of these drugs in vivo, we report the synthesis of 8-fluoropalonosetron (15) that has a binding affinity (Ki = 0.26 ± 0.05 nM) similar to the parent drug (Ki = 0.21 ± 0.03 nM). We radiolabeled 15 by nucleophilic (18)F-fluorination of an unsymmetrical diaryliodonium palonosetron precursor and achieved the radiosynthesis of 1-(methyl-(11)C)-N-granisetron ([(11)C]2) through N-alkylation with [(11)C]CH3I, respectively. Both compounds [(18)F]15 (chemical and radiochemical purity >95%, specific activity 41 GBq/μmol) and [(11)C]2 (chemical and radiochemical purity ≥99%, specific activity 170 GBq/μmol) were evaluated for their utility as positron emission tomography (PET) probes. Using mouse and rat brain slices, in vitro autoradiography with both [(18)F]15 and [(11)C]2 revealed a heterogeneous and displaceable binding in cortical and hippocampal regions that are known to express 5-HT3 receptors at significant levels. Subsequent PET experiments suggested that [(18)F]15 and [(11)C]2 are of limited utility for the PET imaging of brain 5-HT3 receptors in vivo.