EMAIL THIS PAGE TO A FRIEND
Cellular and molecular neurobiology
Piracetam Facilitates the Anti-Amnesic but not Anti-Diabetic Activity of Metformin in Experimentally Induced Type-2 Diabetic Encephalopathic Rats.
PMID 27585927
Abstract
Piracetam exhibits anti-amnesic activity in several animal models of dementia. However, its anti-amnesic potential has yet to be evaluated in type-2 diabetes mellitus (T2DM)-induced encephalopathy. Therefore, in the present study, piracetam (25, 50 and 100 mg/kg) was screened for anti-amnesic and anti-diabetic activity in T2DM-induced encephalopathic male rats. Subsequently, anti-amnesic and anti-diabetic activities were evaluated for piracetam, metformin and their combination in T2DM-induced encephalopathic animals. Rats received streptozotocin (45 mg/kg) and nicotinamide (110 mg/kg) injections on day-1 (D-1) of the experimental schedule and were kept undisturbed for 35 days to exhibit T2DM-induced encephalopathy. All drug treatments were continued from D-7 to D-35 in both experiments. Piracetam (100 mg/kg) attenuated loss in learning and memory in terms of increase in escape latency on D-4 (D-34) and decrease in time spent in the target quadrant on D-5 (D-35) of Morris water maze test protocol, and spatial memory in terms of reduced spontaneous alternation behavior in Y-maze test of encephalopathic rats. Additionally, piracetam attenuated altered levels of fasting plasma glucose and insulin, HOMA-IR and HOMA-B in encephalopathic animals, comparatively lesser than metformin. In the next experiment, combination of piracetam and metformin exhibited better anti-amnesic but not anti-diabetic activity than respective monotherapies in encephalopathic rats. Further, the combination attenuated reduced acetylcholine level and increased acetylcholinesterase activity, increased glycogen synthase kinase-3β level and decreased brain-derived neurotropic factor level in hippocampus and pre-frontal cortex of encephalopathic animals. Thus, piracetam could be used as an adjuvant to metformin in the management of dementia in T2DM-induced encephalopathy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6500
Acetylcholine bromide, ≥99%
C7H16BrNO2