EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Chronic exposure to KATP channel openers results in attenuated glucose sensing in hypothalamic GT1-7 neurons.
PMID 27618741
Abstract
Individuals with Type 1 diabetes (T1D) are often exposed to recurrent episodes of hypoglycaemia. This reduces hormonal and behavioural responses that normally counteract low glucose in order to maintain glucose homeostasis, with altered responsiveness of glucose sensing hypothalamic neurons implicated. Although the molecular mechanisms are unknown, pharmacological studies implicate hypothalamic ATP-sensitive potassium channel (KATP) activity, with KATP openers (KCOs) amplifying, through cell hyperpolarization, the response to hypoglycaemia. Although initial findings, using acute hypothalamic KCO delivery, in rats were promising, chronic exposure to the KCO NN414 worsened the responses to subsequent hypoglycaemic challenge. To investigate this further we used GT1-7xa0cells to explore how NN414 affected glucose-sensing behaviour, the metabolic response of cells to hypoglycaemia and KATP activity. GT1-7xa0cells exposed to 3 or 24xa0h NN414 exhibited an attenuated hyperpolarization to subsequent hypoglycaemic challenge or NN414, which correlated with diminished KATP activity. The reduced sensitivity to hypoglycaemia was apparent 24xa0h after NN414 removal, even though intrinsic KATP activity recovered. The NN414-modified glucose responsiveness was not associated with adaptations in glucose uptake, metabolism or oxidation. KATP inactivation by NN414 was prevented by the concurrent presence of tolbutamide, which maintains KATP closure. Single channel recordings indicate that NN414 alters KATP intrinsic gating inducing a stable closed or inactivated state. These data indicate that exposure of hypothalamic glucose sensing cells to chronic NN414 drives a sustained conformational change to KATP, probably by binding to SUR1, that results in loss of channel sensitivity to intrinsic metabolic factors such as MgADP and small molecule agonists.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0553
NN414, ≥98% (HPLC)
C9H10ClN3O2S2