EMAIL THIS PAGE TO A FRIEND
Fish & shellfish immunology
CpG ODN mimicking CpG rich region of myxosporean Myxobolus supamattayai stimulates innate immunity in Asian sea bass (Lates calcarifer) and defense against Streptococcus iniae.
PMID 27629917
Abstract
Oligodeoxynucleotides (ODNs) containing unmethylated cytosine-phosphate-guanine CpG dinucleotides within specific sequence contexts (CpG motifs) have been reported as pathogen-associated molecular patterns (PAMPs). Its immunostimulatory effects have been demonstrated in diverse vertebrate models. CpG ODN is typically found in bacterial or viral genome and recognized by a non-self recognition receptor Toll-like receptor9 (TLR9). Here, a new CpG ODN 1013 which mimics sequence of SSU rDNA of early eukaryotic organism myxosporidia, Myxobolus supamattayai, was employed to stimulate the immune responses of Asian sea bass Lates calcarifer. Its immunostimulant potentiality was comparatively compared with that of CpG ODN 1668, a widely used as functional immunostimulant. Both unmethylated CpG ODNs with some modified phosphorothioated positions were intraperitoneally injection (5xa0μg/fish). Hematological examination, immunological assays and immune-related genes expression were evaluated 12xa0h, 1, 3 and 5xa0d after post CpG ODN challenge. The immunosimulatory effect of these CpG ODNs on fish immunity to protect the bacterial pathogen Streptococcus iniae was also determined. The results demonstrated that these two CpG ODNs could induce immune responses in Asian sea bass including the significant (Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4875
Nα-Benzoyl-DL-arginine 4-nitroanilide hydrochloride, ≥98%
C19H22N6O4 · HCl