EMAIL THIS PAGE TO A FRIEND
Clinical oral investigations
Oral health and orofacial pain in older people with dementia: a systematic review with focus on dental hard tissues.
PMID 27631597
Abstract
The aim of this review was to provide a systematic overview including a quality assessment of studies about oral health and orofacial pain in older people with dementia, compared to older people without dementia. A systematic literature search was performed in PubMed, CINAHL, and the Cochrane Library. The following search terms were used: dementia and oral health or stomatognathic disease. The quality assessment of the included articles was performed using the Newcastle-Ottawa Scale (NOS). The search yielded 527 articles, of which 37 were included for the quality assessment and quantitative overview. The median NOS score of the included studies was 5, and the mean was 4.9 (SD 2.2). The heterogeneity between the studies was considered too large to perform a meta-analysis. An equivalent prevalence of orofacial pain, number of teeth present, decayed missing filled teeth index, edentulousness percentage, and denture use was found for both groups. However, the presence of caries and retained roots was higher in older people with dementia than in those without. Older people with dementia have worse oral health, with more retained roots and coronal and root caries, when compared to older people without dementia. Little research focused on orofacial pain in older people with dementia. The current state of oral health in older people with dementia could be improved with oral care education of caretakers and regular professional dental care.