EMAIL THIS PAGE TO A FRIEND
Biochemistry international
Large-scale synthesis of the mRNA analogue C17AUGA4C17.
PMID 2764961
Abstract
The large-scale synthesis of the mRNA analogue C17AUGA4C17 is described. This molecule is 41 bases in length and contains three unique codons in its central region. tRNA binding studies at 6 mM Mg2+ in the absence and presence of the labeled heteropolymer show the mutual stimulation of tRNA and mRNA binding and demonstrate that this mRNA provides a simple and suitable system for the analysis of the reactions of the ribosomal elongation cycle.