EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Effect of single and combination of three triazine metabolites at environmental concentrations on early life stages of common carp (Cyprinus carpio L.).
PMID 27650852
Abstract
The sensitivity of early life stages of common carp (Cyprinus carpio L.) to chronic exposure to single and combined environmental concentrations of the triazine metabolites terbuthylazine 2-hydroxy, terbuthylazine-desethyl and atrazine 2-hydroxy was evaluated under laboratory conditions. Their effects were assessed on lipid peroxidation, antioxidant enzymes (total superoxide dismutase, glutathione reductase, catalase, glutathione S-transferase, reduced glutathione), mortality, growth, development and histology. Single metabolites (terbuthylazine 2-hydroxy-0.73xa0μg/L; terbuthylazine-desethyl-1.80xa0μg/L; atrazine 2-hydroxy-0.66xa0μg/L) and combinations were not associated with negative effects on hatching, behaviour, embryo viability, growth or early ontogeny. Carp exposed to terbuthylazine-desethyl at 1.80xa0μg/L showed significantly lower total superoxide dismutase and glutathione reductase activity compared with the control group. Liver histology revealed diffused steatosis associated with the presence of lipid inclusions in hepatic cells in groups exposed to terbuthylazine-desethyl, atrazine 2-hydroxy and the tested combination of metabolites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ALD00112
1,2,3-Triazine, ≥95%
C3H3N3