EMAIL THIS PAGE TO A FRIEND
Glycoconjugate journal
Internalization of a novel, huge lectin from Ibacus novemdentatus (slipper lobster) induces apoptosis of mammalian cancer cells.
PMID 27658397
Abstract
An N-acetyl sugar-binding lectin (termed iNoL) displaying cytotoxic activity against human cancer cells was isolated from the slipper lobster Ibacus novemdentatus (family Scyllaridae). iNoL recognized monosaccharides containing N-acetyl group, and glycoproteins (e.g., BSM) containing oligosaccharides with N-acetyl sugar. iNoL was composed of five subunits (330, 260, 200, 140, and 30xa0kDa), which in turn consisted of 70-, 40-, and 30-kDa polypeptides held together by disulfide bonds. Electron microscopic observations and gel permeation chromatography indicated that iNoL was a huge (500-kDa) molecule and had a polygonal structure under physiological conditions. iNoL displayed cytotoxic (apoptotic) effects against human cancer cell lines MCF7 and T47D (breast), HeLa (ovarian), and Caco2 (colonic), through incorporation (internalization) into cells. The lectin was transported into lysosomes via endosomes. Its cytotoxic effect and incorporation into cells were inhibited by the co-presence of N-acetyl-D-mannosamine (ManNAc). Treatment of HeLa cells with iNoL resulted in DNA fragmentation and chromatin condensation, through activation of caspase-9 and -3. In summary, the novel crustacean lectin iNoL is incorporated into mammalian cancer cells through glycoconjugate interaction, and has cytotoxic (apoptotic) effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8150
D-(+)-Fucose, ≥98%
C6H12O5
M5500
Melibiose, ≥98% (HPLC)
C12H22O11