EMAIL THIS PAGE TO A FRIEND
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Overexpression of IGFBP3 is associated with poor prognosis and tumor metastasis in nasopharyngeal carcinoma.
PMID 27658775
Abstract
Insulin-like growth factor-binding protein-3 (IGFBP3) is an N-linked glycosylated, phosphorylated protein, which has been reported to regulate cancer progression and metastasis. However, the role of IGFBP3 in tumor metastasis remains under debate. Nasopharyngeal carcinoma (NPC) is a highly metastatic head and neck cancer. And it fails to achieve the desired therapeutic efficacy in patients with metastasis, while the role of IGFBP3 in NPC is still unclear. In this study, we first used immunohistochemistry to explore the expression of IGFBP3 in NPC tissues. We found that IGFBP3 was significantly elevated in NPC and its expression level was correlated with N classification, distant metastasis, and TNM clinical stage (all Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU132051 MISSION® esiRNA, esiRNA human IGFBP3 (esiRNA1)