EMAIL THIS PAGE TO A FRIEND
Cancer biomarkers : section A of Disease markers
Detection of survivin, carcinoembryonic antigen and ErbB2 level in oral squamous cell carcinoma patients.
PMID 27662323
Abstract
The aim of this study was to detect the survivin, carcinoembryonic antigen (CEA) and ErbB2 in the saliva, serum and local tumor-exfoliated cells of oral squamous cell carcinoma (OSCC) patients, for providing reliable tumor markers for the early detection of oral malignant cancer. The saliva, serum, and local tumor-exfoliated cell samples of 26 OSCC patients without chemotherapy and 10 non-cancer patients were collected in Department of Oral and Maxillofacial Surgery, School of Stomatology, Peking University. The contents of survivin, CEA and ErbB2 using were detected usingenzyme-linked immunosorbent assay. The survivin and CEA levels in saliva and local tumor-exfoliated cells of OSCC patients were significantly higher than those in the non-cancer patients (P < 0.05), but there was no significant difference in the content of the above factors in the serum sample between two groups. There was no significant difference in the ErbB2 content in the saliva, serum or local tumor-exfoliated cells between two groups. Survivin and CEA levels are significantly increased in the saliva and local tumor-exfoliated cells in OSCC patients, and they can be used as reliable markers for the early detection of oral malignant cancer.