EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Inhibitory Effects of Multiple-Dose Treatment with Baicalein on the Pharmacokinetics of Ciprofloxacin in Rats.
PMID 27671796
Abstract
Ciprofloxacin is used as a treatment for urinary and respiratory tract infections in clinical practice. Baicalein, a major flavonoid present in Scutellaria baicalensis, is a well-known and potent antibacterial compound used in complementary and alternative medicine practices. The present study aimed to clarify the effects of multiple-dose treatment with baicalein on the pharmacokinetics of ciprofloxacin in rats. Following the oral administration of baicalein (20, 40, or 80 mg/kg) for five consecutive days, the rats received an oral administration of ciprofloxacin (20 mg/kg). Blood samples were collected at specific time points, and the plasma concentrations of ciprofloxacin were determined by using high-performance liquid chromatography. To evaluate the mechanisms underlying the interaction between baicalein and ciprofloxacin, a rhodamine 123 accumulation assay was performed in LS-180 cells. A pharmacokinetic study revealed that multiple-dose treatment with baicalein significantly decreased the peak serum concentration (Cmax ), area under the curve (AUC0 → 480 min ), and relative bioavailability (Frel ) of ciprofloxacin (p < 0.05). The rhodamine 123 accumulation assay revealed that treatment with baicalein for 48 h markedly reduced the intracellular accumulation of rhodamine 123. Taken together, these findings suggest that baicalein may result in the therapeutic failure of ciprofloxacin or other quinolone-based antibiotics used for chemotherapy in clinical practice. Copyright © 2016 John Wiley & Sons, Ltd.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
465119
Baicalein, 98%
C15H10O5