EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
MicroRNA-200c modulates DUSP-1 expression in diabetes-induced cardiac hypertrophy.
PMID 27696308
Abstract
Mitogen-activated protein kinases (MAPKs) (ERK1/2, JNK, and p38) are upregulated in diabetic cardiomyopathy (DCM). Dual-specific phosphatase-1 (DUSP-1) has been reported to regulate the activity of MAPKs in cardiac hypertrophy; however, the role of DUSP-1 in regulating MAPKs activity in DCM is not known. MicroRNAs have been reported to regulate the expression of several genes in hypertrophied failing hearts. However, little is known about the microRNAs regulating DUSP-1 expression in diabetes-related cardiac hypertrophy. In the present study, we investigated the role of DUSP-1 and miR-200c in diabetes-induced cardiac hypertrophy. DCM was induced in Wistar rats by low-dose Streptozotocin high-fat diet for 12xa0weeks. Cardiac expression of ERK, p-38, JNK, DUSP-1, miR-200c, and hypertrophy markers (ANP and β-MHC) was studied in DCM in control rats and in high-glucose (HG)-treated rat neonatal cardiomyocytes. miR-200c inhibition was performed to validate DUSP-1 as target. A significant increase in phosphorylated ERK, p38, and JNK was observed in DCM model and in HG-treated cardiomyocytes (pxa0