EMAIL THIS PAGE TO A FRIEND
Journal of applied biomaterials & functional materials
Biopolyester-based systems containing naturally occurring compounds with enhanced thermo-oxidative stability.
PMID 27716869
Abstract
This work presents a sustainable approach for the stabilization of polylactic acid (PLA) against thermo-oxidative aging. Naturally occurring phenolic and polyphenolic compounds, such as ferulic acid (FerAc), vanillic acid (VanAc), quercetin (Querc) and vitamin E (VitE), were introduced into PLA. The preliminary characterization of the systems formulated containing different amounts of natural stabilizers showed that all compounds used acted as plasticizers, leading to a decrease in rheological functions with respect to neat PLA, without significantly modifying the crystallinity of the raw material. The study of the thermo-oxidative behavior of neat PLA and PLA/natural compound systems, performed by spectrometric and thermal analyses, indicated that all stabilizers considered were able to exert a remarkable antioxidant action against thermo-oxidative phenomena. All natural compounds considered are thus proposed as ecofriendly stabilizers, to get fully bio-based polymer systems with enhanced thermo-oxidative stability, suitable for biomedical applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90034
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical reference material
C10H10O4