EMAIL THIS PAGE TO A FRIEND
Chemosphere
Characterization of polyurethane foam (PUF) and sorbent impregnated PUF (SIP) disk passive air samplers for measuring organophosphate flame retardants.
PMID 27723477
Abstract
This study aimed to characterize the uptake of organophosphate esters (OPEs) by polyurethane foam (PUF) and sorbent-impregnated polyurethane foam (SIP) disk passive air samplers (PAS). Atmospheric OPE concentrations were monitored with high-volume active air samplers (HV-AAS) that were co-deployed with passive air samplers. Samples were analyzed for tris(2-chloroisopropyl) phosphate (TCIPP), tri(phenyl) phosphate (TPhP), tris(2-chloroethyl) phosphate (TCEP), and tris(2,3-dichloropropyl) phosphate (TDCIPP). The mean concentration of ∑OPEs in air was 2650xa0pg/m(3) for the HV-AAS. Sampling rates and the passive sampler medium (PSM)-air partition coefficient (KPSM-Air) were calculated for individual OPEs. The average calculated sampling rates (R) for the four OPEs were 3.6xa0±xa01.2 and 4.2xa0±xa02.0xa0m(3)/day for the PUF and SIP disks, respectively, and within the range of the recommended default value of 4xa0±xa02xa0m(3)/day. Since most of the OPEs remained in the linear uptake phase during the study, COSMO-RS solvation theory and an oligomer-based model were used to estimate KPUF-Air for the OPEs. The estimated values of log KPUF-Air were 7.45 (TCIPP), 9.35 (TPhP), 8.44 (TCEP), and 9.67 (TDCIPP). Finally, four configurations of the PUF and SIP disks were tested by adjusting the distance of the gap opening between the upper and lower domes of the sampler housing: i.e. 2xa0cm, 1xa0cm, no gap and 1xa0cm overlap. The sampling rate did not differ significantly between these four configurations (pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
XAD4 Amberlite® XAD4, 20-60 mesh