EMAIL THIS PAGE TO A FRIEND
Metabolomics : Official journal of the Metabolomic Society
Metabolomics reveals dose effects of low-dose chronic exposure to uranium in rats: identification of candidate biomarkers in urine samples.
PMID 27729830
Abstract
Data are sparse about the potential health risks of chronic low-dose contamination of humans by uranium (natural or anthropogenic) in drinking water. Previous studies report some molecular imbalances but no clinical signs due to uranium intake. In a proof-of-principle study, we reported that metabolomics is an appropriate method for addressing this chronic low-dose exposure in a rat model (uranium dose: 40xa0mgxa0L(-1); duration: 9xa0months, nxa0=xa010). In the present study, our aim was to investigate the dose-effect pattern and identify additional potential biomarkers in urine samples. Compared to our previous protocol, we doubled the number of rats per group (nxa0=xa020), added additional sampling time points (3 and 6xa0months) and included several lower doses of natural uranium (doses used: 40, 1.5, 0.15 and 0.015xa0mgxa0L(-1)). LC-MS metabolomics was performed on urine samples and statistical analyses were made with SIMCA-P+ and R packages. The data confirmed our previous results and showed that discrimination was both dose and time related. Uranium exposure was revealed in rats contaminated for 9xa0months at a dose as low as 0.15xa0mgxa0L(-1). Eleven features, including the confidently identified N1-methylnicotinamide, N1-methyl-2-pyridone-5-carboxamide and 4-hydroxyphenylacetylglycine, discriminated control from contaminated rats with a specificity and a sensitivity ranging from 83 to 96xa0%, when combined into a composite score. These findings show promise for the elucidation of underlying radiotoxicologic mechanisms and the design of a diagnostic test to assess exposure in urine, in a dose range experimentally estimated to be above a threshold between 0.015 and 0.15xa0mgxa0L(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7396
(±)-2-Hydroxyoctanoic acid
C8H16O3