EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
A Fluoro-Chromogenic Sensor Based on Organic Molecular Framework for Cu(2+) and F(-) in Aqueous Soluble DMSO.
PMID 27730418
Abstract
The 2,2'-dinaphtholazobenzene molecular framework (P) was designed, synthesized and characterized. Its absorption and fluorescence properties revealed that P is a dual sensor for copper ions (Cu(2+)) and fluoride ions (F(-)) in DMSO. The colorimetric activities were clearly visible by naked eye upon the addition of the two ions. Fluorescence quenching and enhancement were observed when Cu(2+) and F(-) ions were added respectively. Density Functional Theory (DFT) calculations were carried out to provide an insight into the interaction of guest ions (Cu(2+) and F(-)) with P, and to explain how the molecular orbitals were affected. Graphical Abstract ᅟ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS000044
1-Hydroxy-2-naphthaldehyde, AldrichCPR
C11H8O2