EMAIL THIS PAGE TO A FRIEND
Journal of anesthesia
Preventive effects of dexmedetomidine on the development of cognitive dysfunction following systemic inflammation in aged rats.
PMID 27738803
Abstract
In the present study, we examined whether and by what mechanisms dexmedetomidine (DMED) prevents the development of systemic inflammation (SI)-induced cognitive dysfunction in aged rats. Animals received a single intraperitoneal (i.p.) injection of either 5.0xa0mg/kg lipopolysaccharide (LPS) or vehicle. LPS-treated rats were further divided into three groups: early DMED, late DMED, or midazolam (MDZ) treatment (nxa0=xa012 each). Seven days after LPS injection, cognitive function was evaluated using a novel object recognition task, followed by measurement of hippocampal levels of proinflammatory cytokines and Toll-like receptor 4 (TLR-4) expression. For ex vivo experiments, microglia were isolated from the hippocampus for assessment of cytokine response to LPS. LPS-treated rats showed memory deficits, hippocampal neuroinflammation, and TLR-4 upregulation as compared to saline-treated animals. However, early DMED treatment was able to attenuate these SI-induced neurocognitive changes, whereas no benefits were observed in the MDZ and late DMED treatment groups. In ex vivo experiments, early DMED treatment prevented the development of SI-induced excessive microglial hyperactivation, which was blocked by the nonspecific α2-adrenergic receptor (AR) antagonist atipamezole or the specific α2A-AR antagonist BRL-44408, but not by the specific α2B/C-AR antagonist ARC-239. On the other hand, neither DMED nor MDZ had a direct effect on LPS-induced release of pro-inflammatory cytokines from hippocampal microglia at clinically relevant concentrations. Our findings highlight that treatment with DMED during, but not after, peripheral SI can prevent subsequent hippocampal neuroinflammation, overexpression of TLR-4 in microglia, and cognitive dysfunction, as mediated by the α2A-AR signaling pathway.