EMAIL THIS PAGE TO A FRIEND
Oncology reports
Effects of phycoerythrin from Gracilaria lemaneiformis in proliferation and apoptosis of SW480 cells.
PMID 27748904
Abstract
We studied phycoerythrin (PE) in human SW480 tumor cells and the underlying molecular mechanisms of action. PE inhibited cell proliferation as evidenced by CCK-8 assay. The IC50 values of phycoerythrin were 48.2 and 27.4xa0µg/ml for 24 and 48 h of exposure, respectively. PE induced apoptosis and cell cycle arrest in SW480 cells as observed under electron microscopy and with flow cytometry. Apoptosis increased from 5.1 (controls) to 39.0% in 80.0xa0µg/ml PE-treated cells. Differences in protein expression were identified using proteomic techniques. Protein spots (1018±60 and 1010±60) were resolved in PE-treated and untreated group. Forty differential protein spots were analyzed with MALDI-TOF-MS, including GRP78 and NPM1. The expression as measured by qPCR and western blotting agreed with data from two-dimensional electrophoresis. GRP78, NPM1, MTHSP75, Ezrin and Annexinxa0A2 were decreased and HSP60 was increased after PE treatment, indicating that PE may target multiple proteins to induce apoptosis.