EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Resveratrol induces mitochondrial respiration and apoptosis in SW620 colon cancer cells.
PMID 27760368
Abstract
The polyphenol resveratrol (RSV) is found in the skin of red grapes and has been reported to exhibit anticancer properties. The antitumor effects of RSV in the gastrointestinal tract have gained considerable interest due to the high exposure of this tissue to this dietary compound. One of the hallmarks of cancer cells is their particular metabolism mainly relying on glycolysis for ATP production rather than mitochondrial oxidative phosphorylation. Although RSV has been described to act as a calorie-restriction mimetic, modulating energy metabolism in normal tissues, little efforts have been done to study the effects of this polyphenol in the metabolism of cancer cells. Taking this into account, the aim of this study was to explore metabolic effects of this polyphenol in colon cancer. Oxygen consumption, ATP levels, Western blotting and other molecular biology techniques were carried out to characterize the metabolic signature of RSV in SW620 colon cancer cells. Paradoxically, the cytotoxic effects of RSV were associated with an increase in oxygen consumption supported by mitochondrial biogenesis and increased fatty acid oxidation. This partial reversion of the Warburg effect was followed by hyperpolarization of mitochondrial membrane and ROS production, leading to an increased apoptosis. Our results propose that the anticancer mechanisms of RSV could reside in targeting cancer cell metabolism, promoting mitochondrial electron transport chain overload and, ultimately, increasing ROS production. These results shed new light into the anticancer mechanism of RSV supporting the ability of this compound in potentiating the effects of chemotherapy.