EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Impact of Collagen/Heparin Multilayers for Regulating Bone Cellular Functions.
PMID 27762547
Abstract
Bone cell interaction with extracellular matrix (ECM) microenvironment is of critical importance when engineering surface interfaces for bone regeneration. In this work layer-by-layer films of type I collagen (coll), the major constituent of bone ECM, and heparin (hep), a glycosaminoglycan, were assembled on poly(l-lactic acid) (PLLA) substrates to evaluate the impact of the biomacromolecular coating on cell activity. The surface modification of PLLA demonstrated that the hep/coll multilayer is stable after 10 bilayers (confirmed by contact angle, infrared spectroscopy, and morphological analysis). This simple approach provided novel information on the effect of heparin on type I collagen hierarchical organization and subsequent cell response of osteoblast-like (MC3T3-E1) and human bone marrow-derived mesenchymal stem cells (hMSCs). Interestingly, the number of deposited heparin layers (1 or 10) appeared to play an important role in the self-assembly of collagen into fibrils, stabilizing the fibrous collagen layer, and potentially impacting hMSCs activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7653
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
C6H4NNa2O6P